Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700024178.mm.1 |
NM_001163622.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024178.mm.1 |
NM_001163622.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024178.mm.1 |
NM_001163623.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024178.mm.1 |
NM_001163623.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024178.mm.1 |
NM_145984.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024178.mm.1 |
NM_145984.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024178.mm.1 |
TC1700002660.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700024173.mm.1 |
NM_001163622.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024173.mm.1 |
NM_001163622.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024173.mm.1 |
NM_001163623.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024173.mm.1 |
NM_001163623.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024173.mm.1 |
NM_001163624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024173.mm.1 |
NM_001163624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024173.mm.1 |
NM_145984.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024173.mm.1 |
NM_145984.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700024173.mm.1 |
TC1700002660.mm.1 |
MTA TC |