Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159399.3 | NP_001152871.1 | 368 | 52 | ||
GenBank mRNA | DR771512.1 | - | 370 | 52 | ||
DV769287.1 | - | 279 | 52 | |||
JU333020.1 | - | 340 | 52 | |||
JU473760.1 | - | 328 | 52 | |||
JV045812.1 | - | 348 | 52 | |||
JV571796.1 | - | 345 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |