Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sf3b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016441.2 | NP_001016441.1 | 1 | 155 | 69 | |
GenBank mRNA | BC118870.1 | - | 1 | 244 | 69 | |
CR761024.2 | - | 1 | 155 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |