Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115855.4 | NP_191551.1 | 9 - 10 | 1197 | 71 | |
GenBank mRNA | AB005560.1 | - | 9 - 10 | 1024 | 71 | |
AJ001397.1 | - | 9 - 10 | 1099 | 71 | ||
AY081269.1 | - | 9 - 10 | 1110 | 71 | ||
BT000011.1 | - | 9 - 10 | 991 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |