Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129843.3 | NP_181810.2 | 2 - 3 | 325 | 140 | |
GenBank mRNA | AK175397.1 | - | 2 - 3 | 124 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |