Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075072.1 | NP_507473.1 | 3 - 4 | 558 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |