Search
Search

Sec10| Gene Symbol: Sec10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142961.5 | NP_651218.1 | 5 - 6 | 2109 | 67 | |
| GenBank mRNA | AJ242992.1 | - | 5 - 6 | 2084 | 67 | |
| BT150181.1 | - | 5 - 6 | 2107 | 67 | ||
Set Membership: |
|