| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008649.hg.1 |
ENST00000379143.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008649.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008649.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008649.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008649.hg.1 |
TC20000592.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008279.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158370.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158370.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158370.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158370.hg.1 |
NM_182649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158370.hg.1 |
TC2000008279.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000158370.hg.1 |
ENST00000379143.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158370.hg.1 |
ENST00000379160.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158370.hg.1 |
NM_002592.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158370.hg.1 |
NM_182649.1 |
Refseq |