Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000481067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000481067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000481067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000617073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000338591.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000463212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000466300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000481067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000481067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000481067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000617073.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
ENST00000622660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006479.hg.1 |
NM_198317.2 |
Refseq |