| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
232239_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007119.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007119.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007119.hg.1 |
TC10000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007120.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007120.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007120.hg.1 |
TC10000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007121.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007121.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007121.hg.1 |
TC10000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007122.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007122.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007122.hg.1 |
TC10000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007123.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007123.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007123.hg.1 |
TC10000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011389.hg.1 |
ENST00000605714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011389.hg.1 |
ENST00000605714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157077.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157077.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157077.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157077.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157077.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157077.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157077.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157077.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157077.hg.1 |
TC1000008408.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000157082.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157082.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157082.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157082.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157082.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157082.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157082.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157082.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157082.hg.1 |
TC1000008408.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008408.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157085.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157085.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157085.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157085.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157085.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157085.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157085.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157085.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157085.hg.1 |
TC1000008408.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000157076.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157076.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157076.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157076.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157076.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157076.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157076.hg.1 |
TC1000008408.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000157079.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157079.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157079.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157079.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157079.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157079.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157079.hg.1 |
TC1000008408.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000157084.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157084.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157084.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157084.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157084.hg.1 |
ENST00000448490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157084.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157084.hg.1 |
ENST00000448963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157084.hg.1 |
NR_038382.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157084.hg.1 |
TC1000008408.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
232239_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
232239_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232239_at2 |
- |
- |
| Human Genome U95 Set |
61058_PM_at |
- |
- |
| Human Genome U95 Set |
61058_at |
- |
- |
| Human Genome U95 Set |
80843_PM_at |
- |
- |
| Human Genome U95 Set |
80843_at |
- |
- |
| Human Genome U133 Set |
232239_PM_at |
- |
- |
| Human Genome U133 Set |
232239_at |
- |
- |