| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004286.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004286.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004286.hg.1 |
TC10000376.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000389639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000389639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
ENST00000389639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004284.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004284.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004284.hg.1 |
TC10000376.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000389639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000389639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
ENST00000389639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007760.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000152625.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000152625.hg.1 |
ENST00000389639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000152625.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000152625.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000152625.hg.1 |
ENST00000389639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000152625.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000152625.hg.1 |
TC1000007760.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000152627.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000152627.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000152627.hg.1 |
ENST00000330194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000152627.hg.1 |
NM_173554.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000152627.hg.1 |
TC1000007760.hg.1 |
Clariom D TC |