| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
124786 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 8x60K |
46670 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
274342 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
522275 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
225829 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
520655 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
184281 |
AK097020.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
184281 |
BC031333.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
184281 |
BC033095.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
184281 |
NM_003530.3 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
53201 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
350820 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
213453 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
254759 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
137133 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
25000 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
236010 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
349376 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
290542 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
73625 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3328921 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3232032 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_4460 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_4461 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
909 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
13764 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_14_P129180 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3232032 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3328921 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3328921 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3232032 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169768 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169779 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169769 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169784 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626878 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626892 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169780 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169760 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626899 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626886 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169785 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626888 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169776 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169758 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169774 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626877 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626898 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169767 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626884 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626896 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169761 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169769 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169780 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169768 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626877 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169784 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626886 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169758 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169774 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169767 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169776 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169761 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626899 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626884 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626888 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169779 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626892 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626896 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626878 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169785 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09169760 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29626898 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P129180 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16686115 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16686112 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169662 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169777 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16686119 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16686118 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169689 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169747 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169724 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169695 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169735 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169752 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169727 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169712 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169764 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169781 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169669 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169766 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169720 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169749 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169694 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169681 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169788 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169707 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169684 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169761 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169733 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169675 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169673 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169729 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169786 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169668 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169790 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169679 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169769 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169758 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169773 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16686104 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169784 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169742 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169744 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16686108 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169671 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169698 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169779 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16686114 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16686116 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169664 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169701 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169718 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169703 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169755 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169738 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169740 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169774 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169666 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169715 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09169722 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19980238 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P129180 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P40121662 |
- |
- |