Search
Search
| Gene Symbol: Myf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008657.2 | NP_032683.1 | 1 - 2 | 546 | 58 | |
| GenBank mRNA | AK029370.1 | - | 1 - 2 | 562 | 58 | |
| BC119209.1 | - | 1 - 2 | 545 | 8733958 | 58 | |
| BC119211.1 | - | 1 - 2 | 545 | 8733960 | 58 | |
| X59060.1 | - | 1 - 2 | 546 | 58 | ||
Set Membership: |
|