Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173428.3 | NP_775604.3 | 75 - 76 | 11349 | 61 | |
GenBank mRNA | AJ491857.1 | - | 72 - 73 | 10920 | 61 | |
AK165841.1 | - | 11 - 12 | 1708 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |