Search
Search

1700025G04Rik| Gene Symbol: 1700025G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197990.3 | NP_932107.1 | 3 - 4 | 708 | 108 | |
| GenBank mRNA | AI552192.1 | - | 3 - 4 | 357 | 109 | |
| AK148738.1 | - | 2 - 3 | 225 | 108 | ||
| AK167392.1 | - | 359 | 108 | |||
| BC034723.1 | - | 3 - 4 | 487 | 3981517 | 108 | |
| BC055912.1 | - | 3 - 4 | 523 | 3978655 | 108 | |
Set Membership: |
|