| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P133397 |
AY065557.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P133397 |
AK131914.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P133397 |
AY049765.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P133397 |
BC130015.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P133397 |
NM_145126.2 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
267483 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
130648 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
791831 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
267848 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
851808 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
912777 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
516236 |
AK036232.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
516236 |
AK048902.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
516236 |
BC052465.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
516236 |
NM_029758.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
700404 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
810008 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
28475 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
30404 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P143927 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P150609 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P133397 |
AY065557.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P133397 |
AK131914.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P133397 |
AY049765.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P133397 |
BC130015.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P133397 |
NM_145126.2 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401167 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401135 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401168 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401159 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401143 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401154 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401158 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401126 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401173 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401162 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401165 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401160 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401155 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401147 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401182 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401153 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401157 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401145 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22401172 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P150609 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P143927 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P428599 |
AY065557.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P428599 |
AK131914.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P428599 |
AY049765.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P428599 |
BC130015.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P428599 |
NM_145126.2 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133397 |
AY065557.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133397 |
AK131914.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133397 |
AY049765.2 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133397 |
BC130015.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133397 |
NM_145126.2 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P150609 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P143927 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02601907 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02601891 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02601883 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02601881 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401054 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401162 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401112 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401123 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401012 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401051 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401056 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401158 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401058 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401194 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401001 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401026 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401193 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401172 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22400999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401206 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401157 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401120 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401203 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401153 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401055 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401093 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401065 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401199 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401165 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401188 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401043 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401167 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401196 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401062 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401159 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401155 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401215 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401198 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401079 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401210 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401135 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401119 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401010 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401145 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401122 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22401218 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401093 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401160 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401130 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401149 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401168 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401198 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401173 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02601907 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02916942 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401087 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02601887 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401122 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401107 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401104 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02573664 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401123 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02573652 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401154 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401182 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03023288 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401119 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401193 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02601900 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401089 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401120 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401097 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02601893 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401117 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401155 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401167 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401194 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401135 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401111 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401187 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401112 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02601896 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401172 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401171 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401091 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401196 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401158 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03023296 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401153 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02500075 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401190 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401126 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401145 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401143 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22401188 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02868737 |
- |
- |