Search
Search

Notch1| Gene Symbol: Notch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008714.3 | NP_032740.3 | 19 - 20 | 3435 | 77 | |
| GenBank mRNA | AF508809.1 | - | 19 - 20 | 3250 | 77 | |
| AK090118.1 | - | 19 - 20 | 3410 | 77 | ||
| BC138441.1 | - | 19 - 20 | 3250 | 8861461 | 77 | |
| BC138442.1 | - | 19 - 20 | 3250 | 8861462 | 77 | |
| Z11886.1 | - | 19 - 20 | 3249 | 77 | ||
Set Membership: |
|