Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brsk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003920.3 | NP_001003920.2 | 1 - 2 | 292 | 57 | |
NM_001168572.1 | NP_001162044.1 | 1 - 2 | 292 | 57 | ||
GenBank mRNA | AY533671.1 | - | 1 - 2 | 134 | 57 | |
BC086636.1 | - | 1 - 2 | 291 | 6402113 | 57 | |
EU016556.1 | - | 1 - 2 | 140 | 57 | ||
EU016557.1 | - | 1 - 2 | 134 | 57 | ||
EU016558.1 | - | 1 - 2 | 134 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |