| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700017126.mm.1 |
NM_001267583.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017126.mm.1 |
NM_183177.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017126.mm.1 |
TC1700001848.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017127.mm.1 |
NM_001267583.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017127.mm.1 |
NM_183177.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017127.mm.1 |
TC1700001848.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017124.mm.1 |
NM_001267583.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017124.mm.1 |
NM_183177.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017124.mm.1 |
TC1700001848.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017125.mm.1 |
NM_001267583.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017125.mm.1 |
NM_183177.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017125.mm.1 |
TC1700001848.mm.1 |
MTA TC |