Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968065.1 | XP_027823866.1 | 3 - 4 | 363 | 103 | |
XM_027968066.1 | XP_027823867.1 | 3 - 4 | 443 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |