Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fev | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144753.2 | NP_653354.2 | 1 - 2 | 229 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |