Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106656.1 | NP_001100126.1 | 3 - 4 | 440 | 127 | |
XM_006238035.2 | XP_006238097.1 | 3 - 4 | 556 | 127 | ||
XM_006238036.2 | XP_006238098.1 | 3 - 4 | 1475 | 127 | ||
XM_008763697.1 | XP_008761919.1 | 3 - 4 | 556 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |