Search
Search
| Gene Symbol: St7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007639.1 | NP_001007640.1 | 6 - 7 | 1317 | 71 | |
| XM_008761382.1 | XP_008759604.1 | 6 - 7 | 738 | 71 | ||
| GenBank mRNA | BC079194.1 | - | 6 - 7 | 1317 | 7131380 | 71 |
Set Membership: |
|