Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243870.1 | NP_001230799.1 | 6 - 7 | 881 | 70 | |
GenBank mRNA | CF175854.1 | - | 1 - 2 | 177 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |