Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCAGGCCGGGGCCCGGGCCTCCT[C/G]CTCCTCCTCCTCCATCGCCGCCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ARHGEF17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGEF17 - Rho guanine nucleotide exchange factor 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014786.3 | 713 | Missense Mutation | TCC,TGC | S,C 231 | NP_055601.2 | |
XM_017018623.1 | 713 | Intron | XP_016874112.1 | |||
XM_017018624.1 | 713 | Intron | XP_016874113.1 |