Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGATCCAATCACTACTATCACAC[G/T]GGTCTAAGCTTCTACAAAAATCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609642 MIM: 615450 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TARP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TARP - TCR gamma alternate reading frame protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003799.1 | 597 | Intron | NP_001003799.1 | |||
NM_001003806.1 | 597 | Intron | NP_001003806.1 | |||
XM_011515407.2 | 597 | Intron | XP_011513709.1 | |||
XM_017012226.1 | 597 | Intron | XP_016867715.1 | |||
XM_017012227.1 | 597 | Intron | XP_016867716.1 | |||
XM_017012228.1 | 597 | UTR 5 | XP_016867717.1 | |||
XM_017012229.1 | 597 | Intron | XP_016867718.1 | |||
XM_017012230.1 | 597 | Intron | XP_016867719.1 |
TRGC2 - T cell receptor gamma constant 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |