Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTGGATCATAGGCCATGGAGAA[A/T]GTTTGTCCTAAATGTTCCCTTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 187410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNTT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNTT - DNA nucleotidylexotransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OPALIN - oligodendrocytic myelin paranodal and inner loop protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040102.2 | 1467 | UTR 3 | NP_001035191.1 | |||
NM_001040103.2 | 1467 | UTR 3 | NP_001035192.1 | |||
NM_001284320.1 | 1467 | UTR 3 | NP_001271249.1 | |||
NM_001284321.1 | 1467 | UTR 3 | NP_001271250.1 | |||
NM_001284322.1 | 1467 | UTR 3 | NP_001271251.1 | |||
NM_001284323.1 | 1467 | UTR 3 | NP_001271252.1 | |||
NM_001284324.1 | 1467 | UTR 3 | NP_001271253.1 | |||
NM_001284326.1 | 1467 | UTR 3 | NP_001271255.1 | |||
NM_001284327.1 | 1467 | UTR 3 | NP_001271256.1 | |||
NM_033207.4 | 1467 | UTR 3 | NP_149984.1 |