Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACATGGGAACTAGTGCAGAGTGG[C/T]TCATGCCCACGGTTGACAGCCCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609733 | ||||||||||||||||||||
Literature Links: |
LNX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LNX2 - ligand of numb-protein X 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153371.3 | 2156 | Missense Mutation | AAC,AGC | N,S 663 | NP_699202.1 | |
XM_011534995.2 | 2156 | Missense Mutation | AAC,AGC | N,S 540 | XP_011533297.1 | |
XM_017020434.1 | 2156 | Missense Mutation | AAC,AGC | N,S 663 | XP_016875923.1 |