Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTTCTTATATGTTGAATAAGGC[A/G]TGAGGATGTAAGGAAGGATTTCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC101928238 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101928238 - uncharacterized LOC101928238 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF426 - zinc finger protein 426 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300883.2 | 1350 | Missense Mutation | CGC,TGC | R,C 340 | NP_001287812.1 | |
NM_001318055.1 | 1350 | Intron | NP_001304984.1 | |||
NM_001318056.1 | 1350 | Intron | NP_001304985.1 | |||
NM_024106.2 | 1350 | Intron | NP_077011.1 | |||
XM_017027293.1 | 1350 | Missense Mutation | CGC,TGC | R,C 340 | XP_016882782.1 | |
XM_017027294.1 | 1350 | Missense Mutation | CGC,TGC | R,C 136 | XP_016882783.1 |