Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTTTGTTCAGATTGTGTTATA[C/T]GGCAAATTGGTAGAAGCTAGGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604611 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WRN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WRN - Werner syndrome RecQ like helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000553.4 | 4259 | Silent Mutation | TAC,TAT | Y,Y 1157 | NP_000544.2 | |
XM_011544639.2 | 4259 | Silent Mutation | TAC,TAT | Y,Y 1130 | XP_011542941.1 | |
XM_011544640.1 | 4259 | Silent Mutation | TAC,TAT | Y,Y 624 | XP_011542942.1 |