Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCGCTCATCGCACACTGCCGTTT[C/T]TAACATTTTCCGTGGCTTTCTAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNPEP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNPEP - arginyl aminopeptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319182.1 | 110 | UTR 5 | NP_001306111.1 | |||
NM_001319183.1 | 110 | Intron | NP_001306112.1 | |||
NM_001319184.1 | 110 | Intron | NP_001306113.1 | |||
NM_020216.3 | 110 | Intron | NP_064601.3 | |||
XM_005245421.1 | 110 | UTR 5 | XP_005245478.1 | |||
XM_017002022.1 | 110 | Intron | XP_016857511.1 |