Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTGTGCCTAAGGAGCATTTTTT[C/T]CCCAAATGCTTCTACAATACAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611906 MIM: 611889 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
FNDC5 - fibronectin type III domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171940.1 | Intron | NP_001165411.2 | ||||
NM_001171941.2 | Intron | NP_001165412.1 | ||||
NM_153756.2 | Intron | NP_715637.2 |
S100PBP - S100P binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |