Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGTCACTACTTCGGGAAAGCC[A/G]AGGTGCCGTGCCACGGTCCGAGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610976 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOBEC3H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOBEC3H - apolipoprotein B mRNA editing enzyme catalytic subunit 3H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166002.2 | Intron | NP_001159474.2 | ||||
NM_001166003.2 | Intron | NP_001159475.2 | ||||
NM_001166004.2 | Intron | NP_001159476.2 | ||||
NM_181773.4 | Intron | NP_861438.3 | ||||
XM_011529990.2 | Intron | XP_011528292.1 | ||||
XM_011529991.2 | Intron | XP_011528293.1 | ||||
XM_011529992.2 | Intron | XP_011528294.1 |