Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCCTCAGGCTGGACTGGACACC[A/C]AAGAATGGAGCCCACAGAGGAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614092 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RILPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RILPL1 - Rab interacting lysosomal protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319243.1 | Intron | NP_001306172.1 | ||||
NM_001319244.1 | Intron | NP_001306173.1 | ||||
NM_001319302.1 | Intron | NP_001306231.1 | ||||
NM_178314.4 | Intron | NP_847884.2 | ||||
XM_006719372.2 | Intron | XP_006719435.1 | ||||
XM_006719373.3 | Intron | XP_006719436.1 | ||||
XM_011538282.2 | Intron | XP_011536584.1 | ||||
XM_011538284.2 | Intron | XP_011536586.1 | ||||
XM_011538285.2 | Intron | XP_011536587.1 |
SNRNP35 - small nuclear ribonucleoprotein U11/U12 subunit 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |