Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGAACTACTACAGATAGGACAT[G/T]TTTGAGGAAATTTAATCTTTAAGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611754 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AADAT PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
AADAT - aminoadipate aminotransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286682.1 | 1916 | UTR 3 | NP_001273611.1 | |||
NM_001286683.1 | 1916 | UTR 3 | NP_001273612.1 | |||
NM_016228.3 | 1916 | UTR 3 | NP_057312.1 | |||
NM_182662.1 | 1916 | UTR 3 | NP_872603.1 | |||
XM_006714231.2 | 1916 | UTR 3 | XP_006714294.1 | |||
XM_011532020.2 | 1916 | UTR 3 | XP_011530322.1 |
Set Membership: |
HapMap |