Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCAGCTACCTGAGGTGCGAAAC[A/G]GAGTACAATAAGACATTTCGAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102642 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SOAT1 - sterol O-acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252511.1 | Intron | NP_001239440.1 | ||||
NM_001252512.1 | Intron | NP_001239441.1 | ||||
NM_003101.5 | Intron | NP_003092.4 | ||||
XM_011509911.1 | Intron | XP_011508213.1 | ||||
XM_011509912.1 | Intron | XP_011508214.1 |