Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTTCGTGGATGGAGTTGAGGCC[A/G]GGCGGGCCTTTGCCCCCGCAGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615324 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASPDH - aspartate dehydrogenase domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JOSD2 - Josephin domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC4B - leucine rich repeat containing 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080457.1 | 2383 | Silent Mutation | CCC,CCT | P,P 689 | NP_001073926.1 | |
XM_005259429.4 | 2383 | Silent Mutation | CCC,CCT | P,P 689 | XP_005259486.1 | |
XM_006723506.3 | 2383 | Silent Mutation | CCC,CCT | P,P 689 | XP_006723569.1 | |
XM_011527520.2 | 2383 | Silent Mutation | CCC,CCT | P,P 689 | XP_011525822.1 |