Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTATATTTCTTTTAGAAACAGGTT[A/G]TGCTGCTTCCTAATTTGCAGTTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614047 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPGN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPGN - epithelial mitogen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270989.1 | 539 | Intron | NP_001257918.1 | |||
NM_001270990.1 | 539 | Intron | NP_001257919.1 | |||
NM_001270991.1 | 539 | Intron | NP_001257920.1 | |||
NM_001270992.1 | 539 | Intron | NP_001257921.1 | |||
NM_001270993.1 | 539 | Intron | NP_001257922.1 | |||
XM_005265666.4 | 539 | UTR 5 | XP_005265723.1 | |||
XM_011531817.2 | 539 | UTR 5 | XP_011530119.1 |
LOC105377276 - uncharacterized LOC105377276 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTHFD2L - methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |