Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601325 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNTN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNTN3 - contactin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020872.2 | 3128 | UTR 3 | NP_065923.1 | |||
XM_005264757.3 | 3128 | UTR 3 | XP_005264814.1 | |||
XM_011533768.2 | 3128 | UTR 3 | XP_011532070.1 | |||
XM_017006507.1 | 3128 | UTR 3 | XP_016861996.1 | |||
XM_017006508.1 | 3128 | UTR 3 | XP_016861997.1 | |||
XM_017006509.1 | 3128 | UTR 3 | XP_016861998.1 |