Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 000571
          Assay Name: hsa-miR-381
          miRBase Accession Number: MI0000789
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101045920 - 101045994 [+] on Build GRCh38
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Species: Human, Mouse, Bovine, Callithrix jacchus, Cricetulus griseus, Daubentonia madagascariensis, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Rabbit, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 000571
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          UAUACAAGGGCAAGCUCUCUGU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-381-3p MIMAT0000736
          Mouse mmu-miR-381-3p MIMAT0000746
          Bovine bta-miR-381 MIMAT0009307
          Callithrix jacchus cja-miR-381 MIMAT0039446
          Cricetulus griseus cgr-miR-381 MIMAT0023946
          Daubentonia madagascariensis dma-miR-381 MIMAT0049283
          View More Rows
          Dog cfa-miR-381 MIMAT0009891
          Goat chi-miR-381 MIMAT0036199
          Guinea pig cpo-miR-381-3p MIMAT0047226
          Horse eca-miR-381 MIMAT0013149
          Microcebus murinus mmr-miR-381 MIMAT0049192
          Nomascus leucogenys nle-miR-381 MIMAT0049371
          Otolemur garnettii oga-miR-381 MIMAT0049675
          Pan paniscus ppa-miR-381 MIMAT0049040
          Pan troglodytes ptr-miR-381 MIMAT0008133
          Papio hamadryas pha-miR-381 MIMAT0049577
          Pongo pygmaeus ppy-miR-381 MIMAT0015871
          Rabbit ocu-miR-381-3p MIMAT0048426
          Rhesus monkey mml-miR-381-3p MIMAT0006310
          Tupaia chinensis tch-miR-381-3p MIMAT0036568

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-381
          Stem-loop Accession # MI0000789
          Stem-loop Sequence
          UACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGAAUAUACAAGGGCAAGCUCUCUGUGAGUA
          Chromosome Location Chr. 14 - 101045920 - 101045994 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000736
          miRBase ID: hsa-miR-381-3p
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: Chr. 14 - 101045920 - 101045994 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-381
          Stem-loop Accession # MI0000798
          Stem-loop Sequence
          UACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGAAUAUACAAGGGCAAGCUCUCUGUGAGUA
          Chromosome Location Chr. 12 - 109726822 - 109726896 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000746
          miRBase ID: mmu-miR-381-3p
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: Chr. 12 - 109726822 - 109726896 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-381
          Stem-loop Accession # MI0009821
          Stem-loop Sequence
          UACUCAGAGCGAGGUUGCCCUUUGUAUAUUCGGUUUUUUGAUGUAGAAUAUACAAGGGCAAGCUCUCUGUGAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009307
          miRBase ID: bta-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-381
          Stem-loop Accession # MI0031969
          Stem-loop Sequence
          GUUUGGUACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGCAAUAUACAAGGGCAAGCUCUCUGUGAGUAUCAAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039446
          miRBase ID: cja-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-381
          Stem-loop Accession # MI0020517
          Stem-loop Sequence
          UAUUGUUUGGUACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGAAUAUACAAGGGCAAGCUCUCUGUGAGUAUCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023946
          miRBase ID: cgr-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-381
          Stem-loop Accession # MI0040014
          Stem-loop Sequence
          AGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGCAAUAUACAAGGGCAAGCUCUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049283
          miRBase ID: dma-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-381
          Stem-loop Accession # MI0010390
          Stem-loop Sequence
          UACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUAGAAUAUACAAGGGCAAGCUCUCUGUGAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009891
          miRBase ID: cfa-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-381
          Stem-loop Accession # MI0030776
          Stem-loop Sequence
          AGCGAUGGACCUGCCCAAUGCUACUACUGUUUGGUACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUUUUGACGUAGAAUAUACAAGGGCAAGCUCUCUGUGAGUAUCAAACCUUGGACCCAGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036199
          miRBase ID: chi-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-381
          Stem-loop Accession # MI0038761
          Stem-loop Sequence
          AGCGAGGUUGCCCUUUGUAUAUUCGGCUUGUUGACACGGAAUAUACAAGGGCAAGCUCUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047226
          miRBase ID: cpo-miR-381-3p
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-381
          Stem-loop Accession # MI0012894
          Stem-loop Sequence
          UACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUAGAAUAUACAAGGGCAAGCUCUCUGUGAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013149
          miRBase ID: eca-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-381
          Stem-loop Accession # MI0039911
          Stem-loop Sequence
          AGCGAGGUUGCCCUUUGUAUAUUCGGUUUCUUGACGUGCAGUAUACAAGGGCAAGCUCUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049192
          miRBase ID: mmr-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-381
          Stem-loop Accession # MI0040108
          Stem-loop Sequence
          AGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGAAUAUACAAGGGCAAGCUCUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049371
          miRBase ID: nle-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-381
          Stem-loop Accession # MI0040458
          Stem-loop Sequence
          AGCGAGGUUGCCCUUUGUAUAUUCUGUUUAUUGACAUGCAAUAUACAAGGGCAAGCUCUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049675
          miRBase ID: oga-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-381
          Stem-loop Accession # MI0039748
          Stem-loop Sequence
          AGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGAAUAUACAAGGGCAAGCUCUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049040
          miRBase ID: ppa-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-381
          Stem-loop Accession # MI0008654
          Stem-loop Sequence
          ACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGAAUAUACAAGGGCAAGCUCUCUGUGAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008133
          miRBase ID: ptr-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-381
          Stem-loop Accession # MI0040352
          Stem-loop Sequence
          AGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGAAUAUACAAGGGCAAGCUCUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049577
          miRBase ID: pha-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-381
          Stem-loop Accession # MI0014929
          Stem-loop Sequence
          UACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGAAUAUACAAGGGCAAGCUCUCUGUGAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015871
          miRBase ID: ppy-miR-381
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-381
          Stem-loop Accession # MI0039394
          Stem-loop Sequence
          AGCGAGGUUGCCCUGUGUAUAUUCUGUUUAUUGACAUGCAAUAUACAAGGGCAAGCUCUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048426
          miRBase ID: ocu-miR-381-3p
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-381
          Stem-loop Accession # MI0007729
          Stem-loop Sequence
          UACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGAAUAUACAAGGGCAAGCUCUCUGUGAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006310
          miRBase ID: mml-miR-381-3p
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-381
          Stem-loop Accession # MI0031231
          Stem-loop Sequence
          AGCGAGGUUGCCCUUUGUAUGUUCGGUUCAUUGACAUAGAAUAUACAAGGGCAAGCUCUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036568
          miRBase ID: tch-miR-381-3p
          Mature miRNA Sequence: UAUACAAGGGCAAGCUCUCUGU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool A
          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307376_pri
          TaqMan™ Pri-miRNA Assay : Hs03303552_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10242
          Ambion® Pre-miR™ miRNA Precursor : PM10242
          mirVana® miRNA inhibitor : MH10242
          mirVana® miRNA mimic : MC10242
          TaqMan™ Advanced miRNA Assay : mmu482926_mir
          TaqMan™ Advanced miRNA Assay : 477816_mir
          FGMIR : TaqManTM Human MicroRNA Array A
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          FGMIR : TaqManTM Rodent MicroRNA Array A


          miRBase Alias:

          Human: hsa-miR-381 (v18)
          Mouse: mmu-miR-381 (v17)
          Macaca mulatta: mml-miR-381 (v19)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline