Search
Search

| Assay Name: | hsa-miR-381-3p |
|---|---|
| miRBase Accession Number: | MI0000789 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAUACAAGGGCAAGCUCUCUGU |
| Species: | Human, Mouse, Bovine, Callithrix jacchus, Cricetulus griseus, Daubentonia madagascariensis, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Rabbit, Rhesus monkey, Tupaia chinensis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | UAUACAAGGGCAAGCUCUCUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-381-3p | MIMAT0000736 |
| Mouse | mmu-miR-381-3p | MIMAT0000746 |
| Bovine | bta-miR-381 | MIMAT0009307 |
| Callithrix jacchus | cja-miR-381 | MIMAT0039446 |
| Cricetulus griseus | cgr-miR-381 | MIMAT0023946 |
| Daubentonia madagascariensis | dma-miR-381 | MIMAT0049283 |
| View More Rows | ||
| Dog | cfa-miR-381 | MIMAT0009891 |
| Goat | chi-miR-381 | MIMAT0036199 |
| Guinea pig | cpo-miR-381-3p | MIMAT0047226 |
| Horse | eca-miR-381 | MIMAT0013149 |
| Microcebus murinus | mmr-miR-381 | MIMAT0049192 |
| Nomascus leucogenys | nle-miR-381 | MIMAT0049371 |
| Otolemur garnettii | oga-miR-381 | MIMAT0049675 |
| Pan paniscus | ppa-miR-381 | MIMAT0049040 |
| Pan troglodytes | ptr-miR-381 | MIMAT0008133 |
| Papio hamadryas | pha-miR-381 | MIMAT0049577 |
| Pongo pygmaeus | ppy-miR-381 | MIMAT0015871 |
| Rabbit | ocu-miR-381-3p | MIMAT0048426 |
| Rhesus monkey | mml-miR-381-3p | MIMAT0006310 |
| Tupaia chinensis | tch-miR-381-3p | MIMAT0036568 |