Search
Search

| Assay Name: | hsa-miR-146b |
|---|---|
| miRBase Accession Number: | MI0004665 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGAACUGAAUUCCAUAGGCU |
| Species: | Mouse, Alligator mississippiensis, Dog, Horse, Monodelphis domestica, Ornithorhynchus anatinus, Pan troglodytes, Petromyzon marinus, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000103, mir-146 |
Mature miRNA Sequence: | UGAGAACUGAAUUCCAUAGGCU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Mouse | mmu-miR-146b-5p | MIMAT0003475 | |
| Alligator mississippiensis | ami-miR-146b-5p | MIMAT0038172 | |
| Dog | cfa-miR-146b | MIMAT0006667 | |
| Horse | eca-miR-146b-5p | MIMAT0012891 | |
| Monodelphis domestica | mdo-miR-146b-5p | MIMAT0012747 | |
| Ornithorhynchus anatinus | oan-miR-146b-5p | MIMAT0007007 | |
| View More Rows | |||
| Pan troglodytes | ptr-miR-146b | MIMAT0008039 | |
| Petromyzon marinus | pma-miR-146-5p | MIMAT0019478 | |
| Pongo pygmaeus | ppy-miR-146b-5p | MIMAT0015768 | |
| Rhesus monkey | mml-miR-146b-5p | MIMAT0006204 | |