Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10105
          Assay Name: mmu-miR-146b-5p
          miRBase Accession Number: MI0004665
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 46342762 - 46342870 [+] on Build GRCm38
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Species: Mouse, Alligator mississippiensis, Dog, Horse, Monodelphis domestica, Ornithorhynchus anatinus, Pan troglodytes, Petromyzon marinus, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC10105

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000103, mir-146

          Mature miRNA Sequence:

          UGAGAACUGAAUUCCAUAGGCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-146b-5p MIMAT0003475
          Alligator mississippiensis ami-miR-146b-5p MIMAT0038172
          Dog cfa-miR-146b MIMAT0006667
          Horse eca-miR-146b-5p MIMAT0012891
          Monodelphis domestica mdo-miR-146b-5p MIMAT0012747
          Ornithorhynchus anatinus oan-miR-146b-5p MIMAT0007007
          View More Rows
          Pan troglodytes ptr-miR-146b MIMAT0008039
          Petromyzon marinus pma-miR-146-5p MIMAT0019478
          Pongo pygmaeus ppy-miR-146b-5p MIMAT0015768
          Rhesus monkey mml-miR-146b-5p MIMAT0006204

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-146b
          Stem-loop Accession # MI0004665
          Stem-loop Sequence
          GACUGAGAGAACUUUGGCCACCUGGCUCUGAGAACUGAAUUCCAUAGGCUGUGAGCUCUAGCAGACGCCCUAGGGACUCAGUUCUGGUGCCCGGCUGUGCUAUACCAUC
          Chromosome Location Chr. 19 - 46342762 - 46342870 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003475
          miRBase ID: mmu-miR-146b-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Chromosome Location: Chr. 19 - 46342762 - 46342870 [+] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-146b
          Stem-loop Accession # MI0029752
          Stem-loop Sequence
          CGCUCCUUGGCUUUGAGAACUGAAUUCCAUAGGCUUUAAAAGACAAAAAAAGCCCUAUGGAUUCAGUUCUGUAGCUGGGCGGCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038172
          miRBase ID: ami-miR-146b-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-146b
          Stem-loop Accession # MI0008073
          Stem-loop Sequence
          UGAGAACUGAAUUCCAUAGGCUGUGAGCUUGAGCAAACAGCCUAGGGACUCAGUUCUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006667
          miRBase ID: cfa-miR-146b
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-146b
          Stem-loop Accession # MI0012648
          Stem-loop Sequence
          CCUGGCUCUGAGAACUGAAUUCCAUAGGCUGUGAGCUCCAGCAAAUGCCCUAGGGACUCAGUUCUGGUGCCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012891
          miRBase ID: eca-miR-146b-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-146b
          Stem-loop Accession # MI0012503
          Stem-loop Sequence
          CUGGCUCUGAGAACUGAAUUCCAUAGGCUGUGAGCUCAUGCAAAUGCCCUAGGAAGUCAGUUCUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012747
          miRBase ID: mdo-miR-146b-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-146b
          Stem-loop Accession # MI0006788
          Stem-loop Sequence
          AGGCCCAAAUGCGGUGCUUGGCUCUGAGAACUGAAUUCCAUAGGCUGUGAGUUGAUGAGGAUGCCCUAGGGAUUCAGUUCCACAGCCCAGCAACCCACAGCACUGGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007007
          miRBase ID: oan-miR-146b-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-146b
          Stem-loop Accession # MI0008545
          Stem-loop Sequence
          CUGGCACUGAGAACUGAAUUCCAUAGGCUGUGAGCUCUAGCAAUGCCCUGUGGACUCAGUUCUGGUGCCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008039
          miRBase ID: ptr-miR-146b
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Chromosome Location: NA

          Petromyzon marinus

          Stem-loop ID pma-mir-146
          Stem-loop Accession # MI0017081
          Stem-loop Sequence
          CCGGGCGGUGAGAACUGAAUUCCAUAGGCUGUGACGACACUGCAGCAGCCUGGGUCUUCAGUUUUCUCCGUCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019478
          miRBase ID: pma-miR-146-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-146b
          Stem-loop Accession # MI0014835
          Stem-loop Sequence
          CCUGGCACUGAGAACUGAAUUCCAUAGGCUGUGAGCUCUAGCAAAUGCCCUGUGGACUCAGUUCUGGUGCCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015768
          miRBase ID: ppy-miR-146b-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-146b
          Stem-loop Accession # MI0007635
          Stem-loop Sequence
          CCUGGCACUGAGAACUGAAUUCCAUAGGCUGUGAGCUCUAGCAAAUGCCCUAUGGACUCAGUUCUGGUGCCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006204
          miRBase ID: mml-miR-146b-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUAGGCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03308117_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10105
          Ambion® Pre-miR™ miRNA Precursor : PM10105
          TaqMan™ MicroRNA Assay : 001097
          mirVana® miRNA inhibitor : MH10105
          TaqMan™ Advanced miRNA Assay : mmu478513_mir
          TaqMan™ Advanced miRNA Assay : 478513_mir


          miRBase Alias:

          Human: hsa-miR-146b-5p (v21)
          Mouse: mmu-miR-146b (v17)
          Monodelphis domestica: mdo-miR-146b (v19)
          Ochrobactrum anthropi: oan-miR-146b (v18)
          Phocea massiliensis: pma-miR-146 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline