Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002116
          Assay Name: hsa-miR-361-3p
          miRBase Accession Number: MI0000760
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 85903636 - 85903707 [-] on Build GRCh38
          Mature miRNA Sequence: UCCCCCAGGUGUGAUUCUGAUUU
          Species: Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002116
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000172, mir-361

          Mature miRNA Sequence:

          UCCCCCAGGUGUGAUUCUGAUUU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-361-3p MIMAT0004682
          Pan troglodytes ptr-miR-361 MIMAT0008115
          Pongo pygmaeus ppy-miR-361-3p MIMAT0015849
          Rhesus monkey mml-miR-361-3p MIMAT0006287

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-361
          Stem-loop Accession # MI0000760
          Stem-loop Sequence
          GGAGCUUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUCAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUC
          Chromosome Location Chr. X - 85903636 - 85903707 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004682
          miRBase ID: hsa-miR-361-3p
          Mature miRNA Sequence: UCCCCCAGGUGUGAUUCUGAUUU
          Chromosome Location: Chr. X - 85903636 - 85903707 [-] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-361
          Stem-loop Accession # MI0008634
          Stem-loop Sequence
          GGAGCUUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUCAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008115
          miRBase ID: ptr-miR-361
          Mature miRNA Sequence: UCCCCCAGGUGUGAUUCUGAUUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-361
          Stem-loop Accession # MI0014908
          Stem-loop Sequence
          GGAGCUUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUCAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015849
          miRBase ID: ppy-miR-361-3p
          Mature miRNA Sequence: UCCCCCAGGUGUGAUUCUGAUUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-361
          Stem-loop Accession # MI0007707
          Stem-loop Sequence
          GGAGCUUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUCAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006287
          miRBase ID: mml-miR-361-3p
          Mature miRNA Sequence: UCCCCCAGGUGUGAUUCUGAUUU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Human MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303437_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11176
          Ambion® Pre-miR™ miRNA Precursor : PM11176
          mirVana® miRNA inhibitor : MH11176
          mirVana® miRNA mimic : MC11176
          TaqMan™ Advanced miRNA Assay : 478055_mir
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : TaqManTM Human MicroRNA Array B v3
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline