Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002259
          Assay Name: hsa-miR-340*
          miRBase Accession Number: MI0000802
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 180015303 - 180015397 [-] on Build GRCh38
          Mature miRNA Sequence: UCCGUCUCAGUUACUUUAUAGC
          Species: Human, Mouse, Goat
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002259
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000191, mir-340

          Mature miRNA Sequence:

          UCCGUCUCAGUUACUUUAUAGC

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-340-3p MIMAT0000750
          Mouse mmu-miR-340-3p MIMAT0000586
          Goat chi-miR-340-3p MIMAT0036156

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-340
          Stem-loop Accession # MI0000802
          Stem-loop Sequence
          UUGUACCUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUUA
          Chromosome Location Chr. 5 - 180015303 - 180015397 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000750
          miRBase ID: hsa-miR-340-3p
          Mature miRNA Sequence: UCCGUCUCAGUUACUUUAUAGC
          Chromosome Location: Chr. 5 - 180015303 - 180015397 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-340
          Stem-loop Accession # MI0000623
          Stem-loop Sequence
          CAAUUGUACUUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUUA
          Chromosome Location Chr. 11 - 50069702 - 50069799 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000586
          miRBase ID: mmu-miR-340-3p
          Mature miRNA Sequence: UCCGUCUCAGUUACUUUAUAGC
          Chromosome Location: Chr. 11 - 50069702 - 50069799 [+] on Build GRCm38

          Goat

          Stem-loop ID chi-mir-340
          Stem-loop Accession # MI0030751
          Stem-loop Sequence
          GUUUGUACCUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUGUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUUAUACCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036156
          miRBase ID: chi-miR-340-3p
          Mature miRNA Sequence: UCCGUCUCAGUUACUUUAUAGC
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool A
          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Human MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306995_pri
          TaqMan™ Pri-miRNA Assay : Hs03303572_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13080
          Ambion® Pre-miR™ miRNA Precursor : PM13080
          mirVana® miRNA inhibitor : MH13080
          mirVana® miRNA mimic : MC13080
          TaqMan™ Advanced miRNA Assay : 478041_mir
          TaqMan™ Advanced miRNA Assay : mmu482744_mir
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          FGMIR : TaqManTM Rodent MicroRNA Array A
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          FGMIR : TaqManTM Human MicroRNA Array B v3
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0


          miRBase Alias:

          Human: hsa-miR-340* (v17)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline