Search
Search

| Assay Name: | hsa-miR-301a-5p |
|---|---|
| miRBase Accession Number: | MI0000745 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCUCUGACUUUAUUGCACUACU |
| Species: | Human, Mouse, Alligator mississippiensis, Chicken, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Pteropus alecto, Rabbit, Taeniopygia guttata, Xenopus laevis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | GCUCUGACUUUAUUGCACUACU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-301a-5p | MIMAT0022696 | |
| Mouse | mmu-miR-301a-5p | MIMAT0017008 | |
| Alligator mississippiensis | ami-miR-301a-5p | MIMAT0038260 | |
| Chicken | gga-miR-301b-5p | MIMAT0007741 | |
| Columba livia | cli-miR-301a-5p | MIMAT0038611 | |
| Cricetulus griseus | cgr-miR-301a-5p | MIMAT0023890 | |
| View More Rows | |||
| Dasypus novemcinctus | dno-miR-301a-5p | MIMAT0047787 | |
| Guinea pig | cpo-miR-301a-5p | MIMAT0047142 | |
| Pteropus alecto | pal-miR-301a-5p | MIMAT0040045 | |
| Rabbit | ocu-miR-301a-5p | MIMAT0048364 | |
| Taeniopygia guttata | tgu-miR-301-5p | MIMAT0017391 | |
| Xenopus laevis | xla-miR-301-5p | MIMAT0046566 | |