Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC14414
          Assay Name: hsa-miR-301a-5p
          miRBase Accession Number: MI0000745
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 59151136 - 59151221 [-] on Build GRCh38
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Species: Human, Mouse, Alligator mississippiensis, Chicken, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Pteropus alecto, Rabbit, Taeniopygia guttata, Xenopus laevis
          Product Type: mirVana® miRNA mimic
          Assay ID MC14414

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000034, mir-130

          Mature miRNA Sequence:

          GCUCUGACUUUAUUGCACUACU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-301a-5p MIMAT0022696
          Mouse mmu-miR-301a-5p MIMAT0017008
          Alligator mississippiensis ami-miR-301a-5p MIMAT0038260
          Chicken gga-miR-301b-5p MIMAT0007741
          Columba livia cli-miR-301a-5p MIMAT0038611
          Cricetulus griseus cgr-miR-301a-5p MIMAT0023890
          View More Rows
          Dasypus novemcinctus dno-miR-301a-5p MIMAT0047787
          Guinea pig cpo-miR-301a-5p MIMAT0047142
          Pteropus alecto pal-miR-301a-5p MIMAT0040045
          Rabbit ocu-miR-301a-5p MIMAT0048364
          Taeniopygia guttata tgu-miR-301-5p MIMAT0017391
          Xenopus laevis xla-miR-301-5p MIMAT0046566

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-301a
          Stem-loop Accession # MI0000745
          Stem-loop Sequence
          ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGG
          Chromosome Location Chr. 17 - 59151136 - 59151221 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0022696
          miRBase ID: hsa-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: Chr. 17 - 59151136 - 59151221 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-301a
          Stem-loop Accession # MI0000401
          Stem-loop Sequence
          CCUGCUAACGGCUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCGAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGCGAGCAGG
          Chromosome Location Chr. 11 - 87113004 - 87113089 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017008
          miRBase ID: mmu-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: Chr. 11 - 87113004 - 87113089 [+] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-301a
          Stem-loop Accession # MI0029810
          Stem-loop Sequence
          CUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038260
          miRBase ID: ami-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-301b
          Stem-loop Accession # MI0007565
          Stem-loop Sequence
          GUUGCUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUCACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007741
          miRBase ID: gga-miR-301b-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-301a
          Stem-loop Accession # MI0030026
          Stem-loop Sequence
          CUGCUAACGAACGCUCUGACUUUAUUGCACUACUGUACUUCCCAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGAAAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038611
          miRBase ID: cli-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-301a
          Stem-loop Accession # MI0020482
          Stem-loop Sequence
          GCUCGCGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023890
          miRBase ID: cgr-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-301a
          Stem-loop Accession # MI0039058
          Stem-loop Sequence
          GCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047787
          miRBase ID: dno-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-301a
          Stem-loop Accession # MI0038719
          Stem-loop Sequence
          GCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047142
          miRBase ID: cpo-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-301a
          Stem-loop Accession # MI0032465
          Stem-loop Sequence
          GCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040045
          miRBase ID: pal-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-301a
          Stem-loop Accession # MI0039362
          Stem-loop Sequence
          GCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048364
          miRBase ID: ocu-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-301
          Stem-loop Accession # MI0013823
          Stem-loop Sequence
          AGUGCUCUGACUUUAUUGCACUACUGUACUAUACAGAGAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017391
          miRBase ID: tgu-miR-301-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-301-1
          Stem-loop Accession # MI0038315
          Stem-loop Sequence
          GCUCUGACUUUAUUGCACUACUGUACUAUACAGCUAGCAGUGCAAUAGUAUUGUCAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046566
          miRBase ID: xla-miR-301-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA

          Stem-loop ID xla-mir-301-2
          Stem-loop Accession # MI0038316
          Stem-loop Sequence
          GCUCUGACUUUAUUGCACUACUGUACUAUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046566
          miRBase ID: xla-miR-301-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303409_pri
          TaqMan™ Pri-miRNA Assay : Mm03306644_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM14414
          Ambion® Pre-miR™ miRNA Precursor : PM14414
          TaqMan™ MicroRNA Assay : 006346_mat
          mirVana® miRNA inhibitor : MH14414
          TaqMan™ Advanced miRNA Assay : 478796_mir
          TaqMan™ Advanced miRNA Assay : mmu483007_mir


          miRBase Alias:

          Mouse: mmu-miR-301a* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline