Search
Search

| Assay Name: | mmu-miR-365-2* |
|---|---|
| miRBase Accession Number: | MI0001645 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGGACUUUCAGGGGCAGCUGUG |
| Species: | Mouse, Anolis carolinensis, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Ophiophagus hannah, Pteropus alecto, Rabbit |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000061, mir-365 |
Mature miRNA Sequence: | AGGGACUUUCAGGGGCAGCUGUG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Mouse | mmu-miR-365-2-5p | MIMAT0017179 | |
| Anolis carolinensis | aca-miR-365-5p | MIMAT0021947 | |
| Chrysemys picta | cpi-miR-365-5p | MIMAT0037871 | |
| Cricetulus griseus | cgr-miR-365 | MIMAT0041164 | |
| Dasypus novemcinctus | dno-miR-365-2-5p | MIMAT0047837 | |
| Guinea pig | cpo-miR-365-2-5p | MIMAT0047196 | |
| View More Rows | |||
| Ophiophagus hannah | oha-miR-365a-2-5p | MIMAT0036916 | |
| Pteropus alecto | pal-miR-365b-5p | MIMAT0039963 | |
| Rabbit | ocu-miR-365-2-5p | MIMAT0048401 | |