Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481700_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-365-2-5p
          Stem-loop Accession Number: MI0001645
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 79726400 - 79726511 [+] on Build GRCm38
          Mature miRNA Sequence: AGGGACUUUCAGGGGCAGCUGUG
          Species: Mouse, Anolis carolinensis, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Ophiophagus hannah, Pteropus alecto, Rabbit
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481700_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000061, mir-365

          Mature miRNA Sequence:

          AGGGACUUUCAGGGGCAGCUGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-365-2-5p MIMAT0017179
          Anolis carolinensis aca-miR-365-5p MIMAT0021947
          Chrysemys picta cpi-miR-365-5p MIMAT0037871
          Cricetulus griseus cgr-miR-365 MIMAT0041164
          Dasypus novemcinctus dno-miR-365-2-5p MIMAT0047837
          Guinea pig cpo-miR-365-2-5p MIMAT0047196
          View More Rows
          Ophiophagus hannah oha-miR-365a-2-5p MIMAT0036916
          Pteropus alecto pal-miR-365b-5p MIMAT0039963
          Rabbit ocu-miR-365-2-5p MIMAT0048401

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-365-2
          Stem-loop Accession # MI0001645
          Stem-loop Sequence
          AGAGUGAUCAAGGACAGCAAGAAAAAUGAGGGACUUUCAGGGGCAGCUGUGUUUCCUGACUCAGUCAUAAUGCCCCUAAAAAUCCUUAUUGUUCUUGCAGUGUGCAUCGGAG
          Chromosome Location Chr. 11 - 79726400 - 79726511 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017179
          miRBase ID: mmu-miR-365-2-5p
          Mature miRNA Sequence: AGGGACUUUCAGGGGCAGCUGUG
          Chromosome Location: Chr. 11 - 79726400 - 79726511 [+] on Build GRCm38

          Anolis carolinensis

          Stem-loop ID aca-mir-365
          Stem-loop Accession # MI0018861
          Stem-loop Sequence
          AAGAAAAAUGAGGGACUUUCAGGGGCAGCUGUGUUUUACUAACCCAGUCAUAAUGCCCCUAAAAAUCCUUAUUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021947
          miRBase ID: aca-miR-365-5p
          Mature miRNA Sequence: AGGGACUUUCAGGGGCAGCUGUG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-365-2
          Stem-loop Accession # MI0029560
          Stem-loop Sequence
          AGAGGCAGCAAGAAAAAUGAGGGACUUUCAGGGGCAGCUGUGUUUUACUAACCCAGUCAUAAUGCCCCUAAAAAUCCUUAUUGUUCUUGCAAUGUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037871
          miRBase ID: cpi-miR-365-5p
          Mature miRNA Sequence: AGGGACUUUCAGGGGCAGCUGUG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-365-2
          Stem-loop Accession # MI0033094
          Stem-loop Sequence
          CGCAUGUUCAAGGACAGCAAGAAAAAUGAGGGACUUUCAGGGGCAGCUGUGUUUUCUGACUCAGUCAUAAUGCCCCUAAAAAUCCUUAUUGUUCUUGCAGUGUGCAUCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041164
          miRBase ID: cgr-miR-365
          Mature miRNA Sequence: AGGGACUUUCAGGGGCAGCUGUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-365-2
          Stem-loop Accession # MI0039083
          Stem-loop Sequence
          AGGGACUUUCAGGGGCAGCUGUGUUUUCUGACUCAGUCAUAAUGCCCCUAAAAAUCCUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047837
          miRBase ID: dno-miR-365-2-5p
          Mature miRNA Sequence: AGGGACUUUCAGGGGCAGCUGUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-365-2
          Stem-loop Accession # MI0038746
          Stem-loop Sequence
          AGGGACUUUCAGGGGCAGCUGUGUUUUCUGACUCAGUCAUAAUGCCCCUAAAAAUCCUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047196
          miRBase ID: cpo-miR-365-2-5p
          Mature miRNA Sequence: AGGGACUUUCAGGGGCAGCUGUG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-365a-2
          Stem-loop Accession # MI0031465
          Stem-loop Sequence
          UGAGACAGCAAGAAAAAUGAGGGACUUUCAGGGGCAGCUGUGUUUUACUAACCCAGUCAUAAUGCCCCUAAAAAUCCUUAUUGUUCCUUGCAAUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036916
          miRBase ID: oha-miR-365a-2-5p
          Mature miRNA Sequence: AGGGACUUUCAGGGGCAGCUGUG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-365b
          Stem-loop Accession # MI0032423
          Stem-loop Sequence
          AGGGACUUUCAGGGGCAGCUGUGUUUUCUGACUCAGUCAUAAUGCCCCUAAAAAUCCUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039963
          miRBase ID: pal-miR-365b-5p
          Mature miRNA Sequence: AGGGACUUUCAGGGGCAGCUGUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-365-2
          Stem-loop Accession # MI0039381
          Stem-loop Sequence
          AGGGACUUUCAGGGGCAGCUGUGUUUCCUGACUCAGUCAUAAUGCCCCUAAAAAUCCUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048401
          miRBase ID: ocu-miR-365-2-5p
          Mature miRNA Sequence: AGGGACUUUCAGGGGCAGCUGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307505_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19630
          Ambion® Pre-miR™ miRNA Precursor : PM19630
          TaqMan™ MicroRNA Assay : 463537_mat
          mirVana® miRNA inhibitor : MH19630
          mirVana® miRNA mimic : MC19630

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline