Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 464122_mat
          Assay Name: rno-miR-96*
          miRBase Accession Number: MI0000881
          miRBase Version: v22.1
          Chromosome Location: Chr.4: 57463366 - 57463471 [-] on Build Rnor_6.0
          Mature miRNA Sequence: CAAUCAUGUGCAGUGCCAAUAU
          Species: Rat, Dasypus novemcinctus, Guinea pig, Pteropus alecto, Rabbit
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 464122_mat
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000072, mir-96

          Mature miRNA Sequence:

          CAAUCAUGUGCAGUGCCAAUAU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-96-3p MIMAT0017110
          Dasypus novemcinctus dno-miR-96-3p MIMAT0047591
          Guinea pig cpo-miR-96-3p MIMAT0046938
          Pteropus alecto pal-miR-96-3p MIMAT0040135
          Rabbit ocu-miR-96-3p MIMAT0048179

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-96
          Stem-loop Accession # MI0000881
          Stem-loop Sequence
          CCAGUACCAUCUGCUUGGCCGAUUUUGGCACUAGCACAUUUUUGCUUGUGUCUCUCCGCUCUGAGCAAUCAUGUGCAGUGCCAAUAUGGGAAAAGCGGGCUGCUGC
          Chromosome Location Chr. 4 - 57463366 - 57463471 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017110
          miRBase ID: rno-miR-96-3p
          Mature miRNA Sequence: CAAUCAUGUGCAGUGCCAAUAU
          Chromosome Location: Chr. 4 - 57463366 - 57463471 [-] on Build Rnor_6.0

          Dasypus novemcinctus

          Stem-loop ID dno-mir-96
          Stem-loop Accession # MI0038952
          Stem-loop Sequence
          UUUGGCACUAGCACAUUUUUGCUUGUCUCUCUCUGCUCUGAGCAAUCAUGUGCAGUGCCAAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047591
          miRBase ID: dno-miR-96-3p
          Mature miRNA Sequence: CAAUCAUGUGCAGUGCCAAUAU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-96
          Stem-loop Accession # MI0038608
          Stem-loop Sequence
          UUUGGCACUAGCACAUUUUUGCUUGUGUCUCUCUGCUCUGAGCAAUCAUGUGCAGUGCCAAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046938
          miRBase ID: cpo-miR-96-3p
          Mature miRNA Sequence: CAAUCAUGUGCAGUGCCAAUAU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-96
          Stem-loop Accession # MI0032511
          Stem-loop Sequence
          UUUGGCACUAGCACAUUUUUGCUUGUGUCUCUUCGCUCUGAGCAAUCAUGUGCAGUGCCAAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040135
          miRBase ID: pal-miR-96-3p
          Mature miRNA Sequence: CAAUCAUGUGCAGUGCCAAUAU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-96
          Stem-loop Accession # MI0039262
          Stem-loop Sequence
          UUUGGCACUAGCACAUUUUUGCUUGUGUCUCUCCGCUCUGAGCAAUCAUGUGCAGUGCCAAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048179
          miRBase ID: ocu-miR-96-3p
          Mature miRNA Sequence: CAAUCAUGUGCAGUGCCAAUAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464630_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19308
          Ambion® Pre-miR™ miRNA Precursor : PM19308
          mirVana® miRNA inhibitor : MH19308
          mirVana® miRNA mimic : MC19308
          TaqMan™ Advanced miRNA Assay : rno481281_mir


          miRBase Alias:

          Rat: rno-miR-96* (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0